Learn More
Waters Corp XSelect CSH C18 OBD Prep Column, 130Å, 5 µm, 30 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186005423
Encompass_Preferred
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Catalog No. 50-791-434
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.