Learn More
Waters Corp XSelect HSS C18 XP Column, 100Å, 2.5 µm, 2.1 mm X 150 mm, 3/pk
Supplier: Waters Corp 176002897
XSelect HSS C18 is a general purpose, high performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.