Learn More
Waters Corp XSelect HSS C18 Column, 100Å, 3.5 µm, 2.1 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186006380
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection (and lack of endcapping) to produce stability at low pH. The HSS C18 HPLC columns are fully scalable and are completely transferable to UPLC separations.
Catalog No. 50-179-656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.