Learn More
Waters Corp XSelect HSS C18 SB Column, 100Å, 5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186006439
Encompass_Preferred
HSS C18 SB columns are intended for low pH separations that contain complex mixtures of basic and non-basic compounds. HSS C18 SB columns give you alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. HSS C18 SB HPLC columns are fully scalable and transferable to UPLC separations.
Catalog No. 50-179-704
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.