Learn More
Waters Corp XSelect HSS PFP Column, 100Å, 3.5 µm, 4.6 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186005863
Encompass_Preferred
The XSelect HSS PFP is a non-endcapped silica-based HPLC column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. XSelect HSS PFP HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
Catalog No. 50-179-350