Learn More
DWS ADVANTAGE XT30 - 4.6MM X 250MM COLUMN
Supplier:  DWS ADVANTAGE DWNA10649
Encompass
AmberChrom™ Profile™ columns are ideal for the analysis of peptide.They can separate contaminants from the peak of interest and demonstrate slightly different selectivity, as the elution order is somewhat different from the C18 column.AmberChrom™ Profile™ columns are also suitable for separation of proteins or analysis of small molecules and are also an excellent choice for the separation of synthetic oligonucleotides.
Catalog No. NC2481010
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.