Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ XTIRPA™ Kick Plate, Manhole Guard, IX

Catalog No. 19001755 Shop All MSA Products
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-001-755 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-001-755 Supplier MSA™ Supplier No. IN2176
Add to Cart
Add to Cart

MSA XTIRPA confined space entry systems allow for vertical or horizontal entry into many common confined space applications such tanks, vessels, transformers, sewers and manholes. These systems are lightweight, portable and easy to setup allowing for users to save time throughout the workday.

MSA XTIRPA™ Manhole Guard & Manhole Collar Systems

  • Extremely light weight making it easily portable
  • One person set up in just a few minutes
  • Allows for non-entry rescue of a confined space entrant via Workman Rescuer
  • Lift and lower personnel and materials via the Workman Winch eliminating the need for ladders

MSA XTIRPA™ Pole Hoist System

  • Designed for horizontal & vertical confined space entry.

MSA XTIRPA™ Adapter Base System

  • Wide variety of adapter bases for multiple applications
  • Adapter base systems are permanently affixed to existing structures for work areas that require regular set-up and tear-down.
  • Used in conjunction with the MSA XTIRPA universal davit arm for vertical entry into confined spaces.

MSA XTIRPA™ Vehicle Hitch Mount System

  • Customized to accommodate a variety of confined space applications
  • Commonly used with rescue vehicle

MSA XTIRPA™ Counter Weight System

  • Ideal for movement and access between various confined space entry points in the same area

All system components must be purchased separately

Specifications

Material Aluminum
For Use With (Equipment) XTIRPA Systems
Load Bearing Capacity (English) 400 lb.
Load Bearing Capacity (Metric) 181 kg
Weight (English) 4 lb.
Length (English) 52 in.
Product Type Confined Space Entry Equipment

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.