Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XTP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 404000
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-400-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-400-0 Supplier Invitrogen™ Supplier No. 404000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XTP4 Polyclonal Antibody for Western Blot

During the past decade, the role of the ERBB2(neu/HER2) oncogene as an important predictor of patient outcome and response to various therapies in breast cancer has been clearly established. The C35 (C17orf37) is a novel tumor biomarker abundantly expressed in breast cancer. Identification of shared tumor-specific targets is useful in developing broadly applicable therapies. The C35 gene is located on chromosome 17q12, 505nucleotides from the 3' end of the ERBB2oncogene, the antigenic target for trastuzumab(Herceptin) therapy. The chromosomal arrangement of the genes encoding C35 andERBB2 is tail to tail. The ORF 37 open reading frame encodes a 12-kDa protein of unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XTP4
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene MIEN1
Gene Accession No. Q9BRT3
Gene Alias 1810046J19Rik; AI463380; C16H17orf37; C17orf37; C35; HBV XAg-transactivated protein 4; HBV X-transactivated gene 4 protein; MGC14832; MIEN1; Migration and invasion enhancer 1; ORB3; protein C17orf37; protein C17orf37 homolog; Protein C35; RDX12; RGD1306682; XTP4
Gene Symbols MIEN1
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the human C35 protein (chromosome 17 open reading frame 37 (C17orf37 protein), XTP4).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84299
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.