Learn More
Biotium XTT (2,3-Bis-(2-Methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide, disodium salt)
Supplier:  Biotium 10060
Encompass_Preferred
XTT (2,3-Bis-(2-Methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide, disodium salt) is a tetrazolium derivative used widely in cell viability and proliferation testing. The compound is readily reduced in viable cells to a highly water-soluble orange colored product (λ= 475 nm). Applications of the reagent include studies of antifungal susceptibility, drug sensitivity of cells, parasitic nematode viability and tumor cell cytotoxicity. Please also see our XTT Cell Proliferation Assay Kit.Properties: Pale yellow solid, soluble in water; Store desiccated at 4°C; C22H12N7NaO13S2; MW: 675; CAS No. [111072-31-2]
Catalog No. NC9049340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.