Learn More
Abcam XTT Sodium Salt, Tetrazolium Salt, Solid, 100 mg
Description
- Molecular weight: 673.53 g/mol
- Form: Solid
- Application: Used in colorimetric assays for cell viability and proliferation. Cleavage of XTT by dehydrogenase enzymes of metabolically active cells yields a highly colored, water soluble formazan product.
- Advantages: Does not require a formazan crystal solubilization step, unlike MTT.
- Storage: Store at -20°C.
- Shipping: Ambient - Can Ship with Ice.
- Citation count: 2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Dehydrogenase Enzymes |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate | Dehydrogenase enzymes |
| Content And Storage | Store at -20°C. |
| Detection Method | Colorimetric |
| Quantity | 100 mg |
| For Use With (Application) | Colorimetric assays for cell viability and proliferation, valvular injury assessment |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.