Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam XTT Sodium Salt, Tetrazolium Salt, Solid, 25 mg

Catalog No. 502583234
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3234 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3234 Supplier Abcam Supplier No. AB14631025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrazolium salt for colorimetric cell viability and proliferation assays.

  • Molecular weight: 673.53 g/mol
  • Form: Solid
  • Applications: Used in colorimetric assays for cell viability and proliferation. Cleavage of XTT by dehydrogenase enzymes of metabolically active cells yields a highly colored, water-soluble formazan product.
  • Advantages: Does not require a formazan crystal solubilization step unlike MTT assays.
  • Usage examples: Has been used to assess valvular injury for testing cryopreserved cardiac valves.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Dehydrogenase Enzymes
Substrate Dehydrogenase Enzyme Substrate
Content And Storage Store at -20°C.
Detection Location In-Solution Detection
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Detection Method Colorimetric
Quantity 25 mg
For Use With (Application) Colorimetric assays for cell viability and proliferation, valvular injury assessment for cryopreserved cardiac valves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.