Learn More
Abcam Xtt Sodium Salt, Tetrazolium Salt, Solid, 250 mg
Description
- Molecular Weight: 673.53 g/mol
- Applications: Colorimetric assays for cell viability and proliferation
- Performance Characteristics: Cleavage of XTT by dehydrogenase enzymes of metabolically active cells yields a highly colored, water-soluble formazan product, eliminating the need for the formazan crystal solubilization step required by MTT
- Additional Uses: Assessing valvular injury in cryopreserved cardiac valves
- Form: Solid
- Composition: 100% sodium; 4-methoxy-5-[3-(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)tetrazol-3-ium-2-yl]-2-nitrobenzenesulfonate; hydrate
- Storage Temperature: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Dehydrogenase |
| Substrate Properties | Chemical Substrate |
| Substrate | XTT |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Reactivity | Dehydrogenase |
| Shipping Condition | Ambient |
| Detection Method | Colorimetric |
| Quantity | 250 mg |
| For Use With (Application) | Colorimetric assays for cell viability and proliferation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.