Learn More
Abcam XTT Sodium Salt, Tetrazolium Salt, Solid, 50 mg
Description
- Molecular weight: 673.53 g/mol
- Applications: Colorimetric assays for cell viability and cell proliferation
- Form: Solid
- Advantages: Cleavage of XTT by dehydrogenase enzymes in metabolically active cells produces a water-soluble formazan product, eliminating the need for a solubilization step required by MTT
- Usage: Used to assess valvular injury in cryopreserved cardiac valves
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Dehydrogenase |
| Substrate | XTT |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Detection Method | Colorimetric |
| Quantity | 50 mg |
| For Use With (Application) | Colorimetric assays for cell viability and proliferation, assessment of valvular injury in cryopreserved cardiac valves |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.