Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XY 018
GREENER_CHOICE

Catalog No. p-7139266
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-235-0 50 mg
62-231-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-235-0 Supplier Tocris Bioscience™ Supplier No. 6223/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

ROR/NR1F Antagonists

XY 018 is a RORγ antagonist (EC50 = 190 nM). Suppresses androgen receptor expression in prostate cancer cell lines. Also inhibits growth of tumors in androgen receptor-positive xenograft models.

Specifications

CAS 1873358-87-2
Synonym N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide
Quantity 50 mg
Molecular Formula C23H15F7N2O4
Formula Weight 516.36
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.