Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene, MP Biomedicals™

Catalog No. mp215869204 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215869204 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215869204 Supplier MP Biomedicals, Inc Supplier No. 0215869204
Only null left
Add to Cart
Add to Cart

Xylene, MP Biomedicals is a solvent used in Giemsa cell staining protocols.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Synonym Xylol, dimethylbenzene, Xylene mixture of isomers

Specifications

Solubility Information Soluble in absolute alcohol, ether, many organic solvents; practically in insoluble in water.
Formula Weight 106.17
Grade Histological
Quantity 4 L
Physical Form Liquid
Recommended Storage Store at Room Temperature (15-30°C). Protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.