Learn More
Electron Microscopy Sciences Xylene Substitute 5 GL
SDP
Supplier:  Electron Microscopy Sciences 2341205
Encompass_Preferred
An intensive study from 1992 throughout 2000 has shown that Hemo-De (d-Limonene) a terpene-based chemical with a pleasant citrus fragrance, was able to replace many of the clinical laboratories toxic reagents, such as carbol-xylene, xylene, ethyl acetate and formalin.
Hemo-De replaces ethyl acetate in the concentration procedure in parasitology; replaces carbol-xylene, xylene and formalin in the trichrome procedure in parasitology.
Hemo-De replaces xylene in most all procedures in histology. Superior solvent and clearing agent that can be used in all tissue processing, deparaffiniating and slide preparation.
Hemo-De is soluble with alcohol and mounting media. It is biodegradable, non-corrosive, non-flammable (Combustible) and contains no benzene and no toluene. Low toxicity levels. There is minimal tissue shrinkage with reasonably fast drying and leaves no residue.
Catalog No. 50-297-97
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.