Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene (Xylol), Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S02342 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02342 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! : This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.
Catalog No. S02342 Supplier Aldon Corporation Supplier No. IS33000
Add to Cart
Add to Cart
WARNING! : This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.

Chemical Identifiers

CAS 1330-20-7
Molecular Weight (g/mol) 106.17 g/mol

Specifications

Quantity 500 mL
Packaging Amber Glass
Boiling Point 139 to 141°C
Percent Purity 100%
Grade Reagent
Chemical Name or Material Xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.