Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, 99.99%, For ACS analysis, MP Biomedicals™

Catalog No. ICN15869390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15869390 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15869390 Supplier MP Biomedicals, Inc Supplier No. 0215869390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS grade solvent suitable for laboratory applications such as clearing, tissue processing, and general reagent use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
Synonym Xylol

Specifications

Color Colorless
Density 0.84 g/cm3 at 25°C
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 0.9999
Formula Weight 106.17
Grade ACS
Chemical Name or Material Xylenes, ACS, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.