Learn More
Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene)
Description
Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | <-25°C |
| Density | 0.860 |
| Quantity | 4 x 1 L |
| Flash Point | 29°C (84°F) |
| Odor | Aromatic |
| Physical Form | Liquid |
| Refractive Index | 1.497 |
| UN Number | UN1307 |
| Beilstein | 1901563 |
| Merck Index | 14,10081 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H312+H332-H315-H319-H335-H373 |
| Hazard Statement | GHS H Statement H226-H312-H332-H315; Flammable liquid and vapor. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES |
| EINECSNumber | 215-535-7 |
| RTECSNumber | ZE2100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.