Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, ACS Reagent (Dimethylbenzenes), Reagents

Catalog No. 01337630
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-630 4 L
01-337-631 20 L
01-337-629 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-630 Supplier Reagents Holdings Llc Supplier No. C13480004C
Add to Cart
Add to Cart

Xylenes, ACS Reagent (Dimethylbenzenes), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylenes, ACS Reagent (Dimethylbenzenes)
Melting Point -47.9°C to 13.3°C
CAS 1330-20-7
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.87
Molecular Formula C6H4(CH3)2
Synonym Methyl Toluene, Dimethylbenzenes
Boiling Point 137°C to 140°C
Molecular Weight (g/mol) 106.165
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.