Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, Solstice

Catalog No. 60044945 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-945 6 x 1 L Glass Bottle
60-044-941 1 L Glass Bottle
60-044-944 4 x 2.5 L Glass Bottle
60-044-942 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-945 Supplier Solstice Supplier No. 164466X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for analysis (isomeric mixture), >99.0%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Melting Point -48°C
Color Colorless
Density 0.86g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.4960 to 1.4990
Vapor Pressure 18mmHg (37.7°C)
Linear Formula C6H4(CH3)2
UN Number UN1307
Boiling Point 145°C
Molecular Weight (g/mol) 106.16
Formula Weight 106.17g/mol
Grade Reagent
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H304-H312 + H332-H315-H319-H335-H373
DOTInformation class 3 - PG 3 - Xylenes
EINECSNumber 215-535-7
RTECSNumber ZE2100000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.