Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, Purified (Dimethylbenzenes), Reagents

Catalog No. 01336872
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-872 500 mL
01-336-874 20 L
01-336-873 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-872 Supplier Reagents Holdings Llc Supplier No. C2348000500C
Add to Cart
Add to Cart

Xylenes, Purified (Dimethylbenzenes), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym Methyl Toluene, Dimethylbenzenes
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -47.9°C to 13.3°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.87
Boiling Point 137°C to 140°C
Grade Pure
Chemical Name or Material Xylenes, Purified (Dimethylbenzenes)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.