Learn More
AdipoGen Xylenol Blue
SDP
Supplier:  AdipoGen CDXX0006G005
Encompass_Preferred
Chemical. CAS 125-31-5. Formula C23H22O5S. MW 410.48. Synthetic Sulfonephthalein dye. Acid-base pH indicator dye with pH ranges of pH 1.2 Red - pH 2.8 Yellow, and pH 8.0 Yellow - pH 9.6 Blue. Spectral data Absorption, lambdamax 424nm. Has been used as pH sensor, indicator, in cosmetics and drug applications, microbiological assays and determining lipases.
Catalog No. 50-255-9905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.