Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Xylenol Orange, Fluorochrome, 760.6 g/mol, 10 g

Catalog No. 502704765
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4765 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4765 Supplier Abcam Supplier No. AB14627710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorochrome useful for labeling calcifying tissues and detecting lipid hydroperoxide in low density lipoproteins.

  • Molecular Weight: 760.6 g/mol
  • Applications: Useful in single and polychrome sequential labeling of calcifying tissues; detects lipid hydroperoxide in low density lipoproteins by oxidizing ferrous ions (Fe2+ to Fe3+)
  • Detection Method: Spectrophotometry at 560 nm (formation of Fe3+ Xylenol orange complex)
  • Spectral Properties: Absorption λmax 580 nm
  • Composition: 100% Xylenol orange

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Ferrous ions
Target Molecule Lipid Hydroperoxides
Label or Dye Fluorochrome
Buffer 100% Xylenol orange
Content And Storage Store at ambient temperature.
Format Bottle
Detection Method Colorimetric
Formulation 100% Xylenol orange
Quantity 10 g
For Use With (Application) Labeling of calcifying tissues, detection of lipid hydroperoxide in low density lipoproteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.