Learn More
AdipoGen Xylidyl blue I
SDP
Supplier:  AdipoGen CDXX0009G010
Encompass_Preferred
Chemical. CAS 14936-97-1. Formula C25H20N3NaO6S. MW 513.5. Synthetic Colorimetric reagent for Mg detection. The aqueous solution of XB-1 is red, and turns reddish-violet in the presence of Mg at pH 9 maximum wavelength 510 nm, molar absorptivity 49,000, detection range 0.02-0.4 ppm.
Catalog No. 50-255-9901
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.