Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XYLT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548449
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548449 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548449 Supplier Invitrogen™ Supplier No. PA548449
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with mouse and rat based on sequence homology.

Catalyzes the first step in biosynthesis of glycosaminoglycan. Transfers D-xylose from UDP-D-xylose to specific serine residues of the core protein. Initial enzyme in the biosynthesis of chondroitin sulfate and dermatan sulfate proteoglycans in fibroblasts and chondrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XYLT1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Xylt1
Gene Accession No. Q811B1, Q86Y38, Q9EPI1
Gene Alias 8030490L12; beta-D-xylosyltransferase 1; DBQD2; Peptide O-xylosyltransferase 1; PXYLT1; Xt1; XTI; XT-I; Xylosyltransferase 1; xylosyltransferase I; xylosyltransferase iota; XYLT1; XYLT-1; XYLTI; xylT-I
Gene Symbols Xylt1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 126-155 amino acids from the N-terminal region of human XYLT1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233781, 64131, 64133
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.