Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Y 134
GREENER_CHOICE

Catalog No. 267610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-761-0 10 mg
26-765-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-761-0 Supplier Tocris Bioscience™ Supplier No. 2676/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective estrogen receptor modulator (SERM), selective for ERα

Selective estrogen receptor modulator (SERM) that displays selectivity for ERα over ERβ (Ki values are 0.09 and 11.31 nM respectively) and no cross-reactivity with mineralocorticoid, glucocorticoid, androgen and progesterone receptors. Acts as an agonist in the bone and antagonist in reproductive tissue. Suppresses estrogen-stimulated proliferation of ER-positive human breast cancer MCF-7 and T47D cells.

Chemical Identifiers

CAS 849662-80-2
Molecular Formula C28H28N2O3S

Specifications

Quantity 10 mg
Formula Weight 472.6
Purity >98%
Molecular Weight (g/mol) 472.603
Chemical Name or Material Y 134
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.