Learn More
Medchemexpress LLC HY-10583 500mg , Y-27632 (dihydrochloride) CAS:129830-38-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10583500MG
Encompass_Preferred
HY-10583 500mg Y-27632 (dihydrochloride) CAS: 129830-38-2 Y-27632 dihydrochloride is an orally active, ATP-competitive inhibitor of ROCK-I and ROCK-II, with Kis of 220 and 300 nM, respectively. Y-27632 dihydrochloride attenuates Doxorubicin-induced apoptosis of human cardiac stem cells. Y-27632 also suppresses dissociation-induced apoptosis of murine prostate stem/progenitor cells. Y-27632 dihydrochloride primes human induced pluripotent stem cells (hIPSCs) to selectively differentiate towards mesendodermal lineage via epithelial-mesenchymal transition-like modulation[4][5][6][7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0477