Learn More
MilliporeSigma™ Supelco™ Y GlasSeal™ Connector, Fused Silica
Catalog No. 111008993 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
3/Pk.
Catalog No. Packaging Quantity
11-100-8993 3/Pk.
1 options
Catalog No. 11-100-8993 Supplier MilliporeSigma™ Supelco™ Supplier No. 23632
Add to Cart
Add to Cart
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools.
The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
TRUSTED_SUSTAINABILITY
Specifications
Type GC Accessories and Adsorbents
For Use With (Application) Gas Chromatography (GC)
Packaging Quantity 3/Pk.
GlasSeal is a trademark of Sigma-Aldrich Co. LLC