Learn More
Lkt Laboratories CAS 129830-38-2, ≥99%, ROCK inhibitor that prevents binding of Ras-related GTPase Rho A, altering actin cytoskeleton reorganization, cell adhesion, and cell migration.
SDP
Supplier:  Lkt Laboratories Y1000
Encompass_Preferred
Y27632 is a selective inhibitor of Rho-associated protein kinase (ROCK); Y27632 binds to the catalytic site of ROCK, preventing binding of Ras-related GTPase Rho A, which is involved in actin cytoskeleton reorganization, cell adhesion, and cell migration. Y27632 inhibits downstream endothelin and TGF-β-related signaling. This compound exhibits neuroprotective and neuromodulatory activity, displaying benefit in animal models of Parkinson’s Disease and inhibiting conditioned place aversion in animal models of memory formation. Reduces dissociation-induced apoptosis in embryonic and neural stem cells. Increases stem cell cloning efficiency.
Catalog No. NC2539832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.