Learn More
Enzo Life Sciences Y27632 . dihydrochloride, (1mg), CAS Number: 129830-38-2
SDP
Supplier:  Enzo Life Sciences ALX270333M001
Encompass_Preferred
ROCK inhibitor. Y-27632 is a highly potent, cell permeable, selective and ATP competitive inhibitor of ROCK1 and ROCK2 (IC. .There has been a growing interest in Y-27632 for use in stem cell self-renewal and reprogramming. It is known to increase the survival rate of human embryonic stem cells undergoing cryopreservation. Alternative Name: (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide . 2HCl. Formula: C14H21N3O . 2HCl. MW: 247.3 . 73.0. CAS Number: 129830-38-2. Purity: ≥98% (HPLC (UV)). Appearance: White to pale yellow powder. Solubility: Soluble in DMSO (25mg/ml), methanol (25mg/ml), acetonitrile (<1mg/ml), dioxan (<1mg/ml), water (very soluble). Use/Stability: Stock solutions are stable for up to 1 month when stored at -20°C. Long Term Storage: -20°C
Catalog No. NC9527632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.