Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont Industrial™ Y5080 String Knit Glove

Catalog No. 18064011 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Wells Lamont Industrial™ String Knit Glove
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-064-011 Wells Lamont Industrial™ String Knit Glove
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-064-011 Supplier Wells Lamont Industrial™ Supplier No. Y5080L
Add to Cart
Add to Cart

Y5080 string knit glove. Polyester and cotton blend. Ambidextrous.

A polyester and cotton string glove is an ideal everyday general work glove.

  • The blend of the fibers provide comfort to the users hand, the gloves can be laundered for extended wear.
  • Y5080 is an ideal glove for general maintenance, construction, and warehousing.
  • If you need thermal protection, consider the Y1451.

Specifications

ANSI Abrasion Rating ANSI 0
Chlorinated No
Construction Seamless Knit
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A1
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Material Polycotton
Powdered Powder Free
Product Type General Purpose Gloves
Size Large
Sterility Non-sterile
EN 421 No
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.