Learn More
STEMCELL Technologies Verteporfin, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 1000261
Encompass
Verteporfin is a benzoporphyrin derivative that inhibits autophagosome formation by directly targeting and modifying p62, a scaffold protein that targets polyubiquitinated proteins for degradation (Donohue et al.). Verteporfin also disrupts YAP-TEAD interactions by enhancing trypsin cleavage of YAP (EC₅₀ = 100 nM; Brodowska et al.; Liu-Chittenden et al.).CANCER RESEARCH· Inhibits the growth, proliferation, and viability of human retinoblastoma cells (Brodowska et al.).· Inhibits self-renewal in cancer stem cells with high YAP1 and ALDH1 levels (Song et al.).
Catalog No. 50-204-5505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.