Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YBX2 Mouse anti-Human, Mouse, Clone: OTI3C12, liquid, TrueMAB™
SDP

Catalog No. 501683934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3934 Supplier OriGene Supplier No. TA811247
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.

Specifications

Antigen YBX2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene YBX2
Gene Accession No. Q9Y2T7, Q9Z2C8
Gene Alias CONTRIN, CSDA3, DBPC, MSY2
Gene Symbols YBX2
Host Species Mouse
Immunogen Full length human recombit protein of human YBX2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51087, 53422
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.