Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yeast Extract, Powder, Spectrum™ Chemical
SDP

Catalog No. 18614704 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
12 kg
45 kg
Packaging Type:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
18-614-704 100 g Amber Glass Bottle
18-614-705 12 kg Poly Pail
18-614-706 45 kg Fiber Drum
18-614-707 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-614-704 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. YE105100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Specifications

Form Powder
Grade Reagent
Media Type Yeast Extract
Product Type Yeast Media
Packaging Type Amber Glass Bottle
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.