Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Yeast Kex-2 Recombinant Protein, PeproTech®

Catalog No. 4504550UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-45-50UG 50 μg
45045250UG 250 μg
45045500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 450-45-50UG Supplier Gibco™ Supplier No. 4504550UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

450-45-1MG will be provided as 2 x 500 μg (450-45-500UG). Recombinant Yeast Kex-2 is a 60.4 kDa protease consisting of 558 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Kex-2 cleaves at the carboxyl end of the recognition sequences Arg-Arg/X and Lys-Arg/X.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13134
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 855483
Molecular Weight (g/mol) 60.4 kDa
Name Yeast Kex-2
Quantity 50 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Kexin
Common Name KEX2
Gene Symbol KEX2
Biological Activity Recombinant Kex-2 from High-5 insect cells contains the same specific activity and recognition sequence specificity as yeast derived KEX-2. 1 milligram of recombinant KEX-2 contains activity equivalent to at least 40 units of yeast derived KEX-2. Cleaves (at the carboxyl side of K/R-R.)
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPVPAPPMDS SLLPVKEAED KLSINDPLFE RQWHLVNPSF PGSDINVLDL WYNNITGAGV VAAIVDDGLD YENEDLKDNF CAEGSWDFND NTNLPKPRLS DDYHGTRCAG EIAAKKGNNF CGVGVGYNAK ISGIRILSGD ITTEDEAASL IYGLDVNDIY SCSWGPADDG RHLQGPSDLV KKALVKGVTE GRDSKGAIYV FASGNGGTRG DNCNYDGYTN SIYSITIGAI DHKDLHPPYS EGCSAVMAVT YSSGSGEYIH SSDINGRCSN SHGGTSAAAP LAAGVYTLLL EANPNLTWRD VQYLSILSAV GLEKNADGDW RDSAMGKKYS HRYGFGKIDA HKLIEMSKTW ENVNAQTWFY LPTLYVSQST NSTEETLESV ITISEKSLQD ANFKRIEHVT VTVDIDTEIR GTTTVDLISP AGIISNLGVV RPRDVSSEGF KDWTFMSVAH WGENGVGDWK IKVKTTENGH RIDFHSWRLK LFGESIDSSK TETFVFGNDK EEVEPAATES TVSQYSASST SISISATSTS SISIGVETSA IPQTTTASTD PDSDPNTP
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.