Learn More
Research Products International Corp Yeast Nitrogen Base without Amino Acids, 500 Grams
Supplier: Research Products International Corp Y20040500.0
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivaiton of yeasts. Excludes histidine methionine, tryptophan and carbon source. Also contains <1 mg/L each of vitamins and trace inorganic salts.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.