Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Yeast-Torula

For biological research purposes.

$363.00 - $913.00

Specifications

Format Solid
Product Type Yeast-Torula, 
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ23551Q5 View Documents Affymetrix/USB™
J23551Q5
100 lb. Each for $913.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ23551Q1 View Documents Affymetrix/USB™
J23551Q1
25 lb. Each for $363.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Format: Solid
Specifications

Specifications

Solid
Yeast-Torula, 
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit