Learn More
Enzo Life Sciences YEEI peptide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP3000001
Encompass_Preferred
Sequence is from hamster polyoma virus middle T inclusive of Tyr324. This 11-residue phosphopeptide activates Src family members by binding with high affinity (Kd=3-6 nM for Src SH2) to their SH2 domains. The crystal structures of this peptide complexed to Src and Lck SH2 domains have been determined. Purity: ≥98%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1332
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.