Learn More
Checkers Yellow Jacket™ AMS Channel Center 2.25 in. Max Diameter YJ225-AMSC Checkers
Catalog No. 19088401 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-088-401 Black on Yellow
1 options
Catalog No. 19-088-401 Supplier Checkers Supplier No. YJ3225AMSCTRYB
Only null left
Add to Cart
Add to Cart
3-Channel Heavy-Duty Yellow Jacket™ AMS™ System Center Section. The YELLOW JACKET™ 3-CHANNEL AMS™ (Advanced Modular System) is patented technology that utilizes separate cable protector sections and ramps to create a system that is fully expandable to hold an unlimited number of cables.
  • AMS Ramps Sold Individually
  • AMS Female Ramp = YJ3-225-AMS-F-B
  • AMS Male Ramp - YJ3-225-AMS-M-B
Specifications
Color Black on Yellow
Material Polyurethane
Product Type Cable Protector