Learn More
Description
- Ideal for remote places and emergencies
- Safe, non-toxic, and self-contained
- Waterproof, unaffected by wind or weather
- To activate 6 in. lightstick: bend, listen for snap, and shake
- Designed so base will stand without support
- Features top hole to attach string or tie (not included)
Specifications
Specifications
| Lighting Life | 12 hr. |
| Color | Yellow |
| For Use With (Application) | Emergency Light |
| Length (English) | 6 in. |
| Mounting | String or Tie (Not included) |
| Product Type | Lightstick |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.