Learn More
Yellow-Seven (TM) Buffer Solution, pH 7.00 (+/-0.01 at 25°C), Color-coded Yellow, Reference Standard, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Yellow-Seven (TM) Buffer Solution, pH 7.00 (±0.01 at 25°C), Color-coded Yellow, Reference Standard is an inorganic solution in water. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum's own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications
Specifications
| CAS | 7732-18-5, 7778-77-0, 3051-11-4, 1310-73-2 |
| CAS Max % | 99.100%, 0.680%, 0.100%, 0.116% |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 7.00 ±0.01 |
| Physical Form | Aqueous Solution |
| Quantity | 6 x 500 mL |
| Solution Type | pH Buffer |
| Packaging | Poly Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.