Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YES1 Monoclonal Antibody (1612CT505.7.50)
GREENER_CHOICE

Catalog No. PIMA537696
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537696 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537696 Supplier Invitrogen™ Supplier No. MA537696
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YES1 Monoclonal Antibody for Western Blot

YES1 is a recombinant human protein kinase that is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YES1
Applications Western Blot
Classification Monoclonal
Clone 1612CT505.7.50
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Yes1
Gene Accession No. P07947
Gene Alias cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase
Gene Symbols Yes1
Host Species Mouse
Immunogen recombinant protein of human YES1.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.