Learn More
Apexbio Technology LLC ZLN024 hydrochloride 1883548-91-1 5mg
SDP
Supplier:  Apexbio Technology LLC B48405
Encompass_Preferred
ZLN024 hydrochloride (CAS 1883548-91-1) is a small-molecule allosteric activator targeting AMP-activated protein kinase (AMPK) It is designed to selectively modulate heterotrimeric AMPK complexes thereby enhancing enzyme catalytic activity and influencing cellular metabolic pathways ZLN024 hydrochloride exerts its biological activity primarily through allosteric activation of AMPK The compound demonstrates activation with EC50 values of 0 42 M (recombinant AMPK 2 1 1) and ranges from 0 13 M to 1 1 M across various isoforms in vitro Based on these pharmacological properties ZLN024 hydrochloride holds research potential in the investigation of metabolic enzyme regulation and the study of cellular metabolic pathways and disease mechanisms
Catalog No. 50-000-91178
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.