Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YLPM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566368 Supplier Invitrogen™ Supplier No. PA566368
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGNKEPLADTS SNQQKNFKMQ SAAFSIAADV KDVKAAQSNE NLSDSQQEPP KSEVSEGPVE PSNWDQNVQS METQIDKAQA VTQPVPLANK PVPAQST Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.

Plays a role in the reduction of telomerase activity during differentiation of embryonic stem cells by binding to the core promoter of TERT and controlling its down-regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YLPM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YLPM1
Gene Accession No. P49750
Gene Alias C14orf170; Nuclear protein ZAP3; PPP1R169; protein phosphatase 1, regulatory subunit 169; YLP motif containing 1; YLP motif-containing protein 1; YLPM1; ZAP; ZAP113; ZAP3
Gene Symbols YLPM1
Host Species Rabbit
Immunogen Recombinant Human YLPM1. Recombinant protein control fragment (Product #RP-107044).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56252
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.