Learn More
Invitrogen™ YLPM1 Polyclonal Antibody

Description
Immunogen sequence: RDRGELRIRE YPERGDTWRE KRDYVPDRMD WERERLSDRW YPSDVDRHSP MAEHMPSSHH SSEMMGSDAS LDSDQGLGGV MVLSQRQHEI ILKA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
Specifications
Specifications
| Antigen | YLPM1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | YLPM1 |
| Gene Accession No. | P49750 |
| Gene Alias | C14orf170; Nuclear protein ZAP3; PPP1R169; protein phosphatase 1, regulatory subunit 169; YLP motif containing 1; YLP motif-containing protein 1; YLPM1; ZAP; ZAP113; ZAP3 |
| Gene Symbols | YLPM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.