Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam YM-01, Allosteric Hsp70 Modulator, Solid, 10 mg
Description
- Purity: >98%
- Molecular Weight: 417.97 g/mol
- Chemical Class: Allosteric Hsp70 modulator and MKT-077 analog
- Biological Activity: Potent anti-tau agent; promotes triage functions of Hsp70; rapidly and potently reduces tau levels ex vivo; shows antitumor effects in vivo
- Form: Solid
- CAS Number: 409086-68-6
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Hsp70, Tau |
| Alternate Part Number | YM-1 |
| Buffer | 100% YM-01 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% YM-01 |
| Quantity | 10 mg |
| For Use With (Application) | Anti-tau, Antitumor |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?