Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam YM-01, Allosteric Hsp70 Modulator, Solid, 25 mg

Catalog No. 502704630
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4630 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4630 Supplier Abcam Supplier No. AB14642325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent anti-tau agent and allosteric Hsp70 modulator that shows antitumor effects in vivo.

  • Purity: >98%
  • Molecular Weight: 417.97 g/mol
  • Chemical Class: Allosteric Hsp70 modulator; MKT-077 analog
  • Biological Activity: Potent anti-tau agent; promotes triage functions of Hsp70; rapidly and potently reduces tau levels ex vivo; shows antitumor effects in vivo
  • Form: Solid
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Alternate Part Number YM-01
Form Solid
Shipping Condition Ambient
Target Molecule Hsp70
Quantity 25 mg
For Use With (Application) Hsp70 modulation, anti-tau research, antitumor studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.