Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ YM 244769
GREENER_CHOICE

Catalog No. p-4816529
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-441-0 10 mg
45-445-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-441-0 Supplier Tocris Bioscience™ Supplier No. 4544/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Na+/Ca2+ exchange inhibitor (reverse mode); neuroprotective

Inhibitor of the reverse mode of Na+/Ca2+ exchange (NCX). Potently inhibits Na+-dependent 45Ca2+ uptake (IC50 values are 18, 68 and 96 nM for CCL39 cells transfected with NCX3, NCX1 and NCX2 respectively). Displays selectivity for NCX over other Na+ and Ca2+ transporters. Protects against hypoxia/reoxygenation-induced cell damage in neuronal SH-SY5Y cells expressing NCX1 and NCX3.

Chemical Identifiers

CAS 1780390-65-9
Molecular Formula C26H23ClFN3O3

Specifications

Quantity 10 mg
Formula Weight 516.39
Purity >99%
Molecular Weight (g/mol) 479.936
Chemical Name or Material YM 244769
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.