Learn More
Selleck Chemical LLC YM-58483 (BTP2) 5mg 223499-30-7
SDP
Supplier:  Selleck Chemical LLC S83805MG
Encompass_Preferred
YM-58483/BTP2 is a blocker of store-operated Ca2+ entry (SOCE), which regulates the activation of non-excitable cells such as lymphocytes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6011
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.